MilliporeSigma
  • Potential role of the flavin-containing monooxygenases in the metabolism of endogenous compounds.
Potential role of the flavin-containing monooxygenases in the metabolism of endogenous compounds.
Chemico-biological interactions (1995-04-28)
A A Elfarra
ABSTRACT
Several xenobiotics and their corresponding cysteine S-conjugates are metabolized in vivo to cysteine S-conjugate sulfoxides and/or N-acetylcysteine S-conjugate sulfoxides. Homocysteine S-conjugates, such as methionine and ethionine, are also metabolized in vivo to sulfoxides. The enzymatic basis for these metabolic reactions is not known. Recently, the rat liver and kidney S-benzyl-L-cysteine S-oxidase activities were found to be associated with flavin-containing monooxygenases that are structurally and immunochemically related to known FMO1 isoforms. Further evidence for FMO1 being the major FMO isoform involved in S-benzyl-L-cysteine sulfoxidation was obtained from kinetic studies with cDNA-expressed rabbit FMOs. Endogenous cysteine S-conjugates, e.g. cysteinylcatecholamines, cysteinylleukotrienes, lanthionine and djenkolic acid may also be substrates for FMOs, since S-benzyl-L-cysteine can be considered a model for these compounds. Methionine, an endogenous homocysteine S-conjugate, was shown to be a substrate for cDNA-expressed rabbit FMO1, FMO2, and FMO3, however, the methionine sulfoxidation reaction was preferentially catalyzed by FMO3. These results suggest that FMOs may also play a role in the in vivo metabolism of endogenous homocysteine S-conjugates.
Sigma-Aldrich
S-Benzyl-L-cysteine, 97%