MilliporeSigma
  • Membrane protein alterations in rodent erythrocytes and synaptosomes due to aging and hyperoxia.
Membrane protein alterations in rodent erythrocytes and synaptosomes due to aging and hyperoxia.
Biochimica et biophysica acta (1995-04-24)
K Hensley, B J Howard, J M Carney, D A Butterfield
ABSTRACT
We have applied the technique of electron paramagnetic resonance (EPR) protein-specific spin labeling to the study of membrane protein alterations occurring during age and exposure to isobaric hyperoxia. Cortical synaptosomes and erythrocyte membranes (ghosts) were isolated from young rodents (Fisher 344 rats or mongolian gerbils, 3-4 months of age) and aged rodents (age 22-27 months for rats, greater than 15 months for gerbils). Membrane proteins were spin labeled with the thiol-specific spin label MAL-6 (2,2,6,6,-tetramehtyl-4-maleimido-piperdin-1-oxyl). The relevant EPR spectral parameter of MAL-6 labeled membranes, the W/S ratio, decreased significantly with age of animal in both synaptosomes and ghosts (P < 0.001). As a paradigm for accelerated oxidative stress, young and aged gerbils were exposed to an atmosphere of 90-100% O2 for 0-48 h. In both young and aged gerbils, the W/S ratio decreased significantly with hyperoxic stress (P < 0.003). The W/S ratio of synaptosomes isolated from aged gerbils decreased continually from 0-48 h hyperoxia, whereas the W/S ratio of synaptosomes from young animals demonstrated a pronounced rebound effect from 24-48 h. The results are discussed with reference to membrane protein oxidation in aging.