MilliporeSigma
  • The feasibility of early second trimester pregnancy (12 to 14 weeks) termination with single intra-amniotic injection of 15-methyl PGF2 alpha.
The feasibility of early second trimester pregnancy (12 to 14 weeks) termination with single intra-amniotic injection of 15-methyl PGF2 alpha.
The Australian & New Zealand journal of obstetrics & gynaecology (1995-02-01)
R N Prasad, P G Adaikan, C Anandakumar, S S Ratnam
ABSTRACT
Prostaglandin analogues have proven to be safe and effective for second trimester pregnancy terminations when given by various routes. Of these, the intra-amniotic (IA) route has proven to be safer, especially in women with medical disorders, as it causes the least occurrence of systemic side-effects. Previously, blind transabdominal IA instillation of the analogue 15-methyl PGF2 alpha was routinely possible only after 16 weeks' gestation. With the advent of ultrasound-guided techniques, it has now become possible to use the IA route for very early second trimester pregnancies. This study examines the use of single 1.5 mg 15-methyl PGF2 alpha IA injection for termination of 12-14 weeks pregnancies. Single injection resulted in successful abortion in 76.5% of women and, in the unsuccessful cases, additional prostaglandin injection resulted in successful abortion in all women within 48 hours without problems. It is therefore feasible to use ultrasound-guided IA prostaglandin injection for successful termination of early second trimester pregnancies.