MilliporeSigma
Inhibition of plasminogen activation by polymerized ampicillin.
Thrombosis research (1994-09-15)
A al-Roof Higazi, M Hermoni, R Aziza, M Mayer
ABSTRACT
The polymerized beta-lactam antibiotic ampicillin inhibits the proteolytic activity of human plasmin upon 125I-labeled fibrin clots. The inhibition is dose-dependent, with half-maximal inhibition occurring at 1.25 mM of the polymerized antibiotic. Polymerized ampicillin also inhibits binding of plasmin to fibrin, and 38% inhibition of binding occurs at 10 mM of the antibiotic. Furthermore, polymerized ampicillin inhibits the activation of plasminogen by either urokinase-like plasminogen activator (uPA) or tissue type-plasminogen activator (tPA). At 7.5 mM of polymerized ampicillin, the uPA-mediated plasminogen activation is suppressed by 94%, and half-maximal inhibition is obtained at 0.66 mM. The direct activity of uPA on the chromogenic substrate L-pyroglutamyl-glycyl-L-arginine p-nitroanilide hydrochloride (S-2444) is unaffected by polymerized ampicillin levels of up to 10 mM. The inhibitory effects of the polymerized antibiotic on the activation of plasminogen by both uPA and tPA is totally abolished in presence of fibrin. These interactions may serve as a novel model for ligands that enhance the clot-specificity of thrombolytic agents.