MilliporeSigma
  • Discriminative stimulus effects of alpidem, a new imidazopyridine anxiolytic.
Discriminative stimulus effects of alpidem, a new imidazopyridine anxiolytic.
Psychopharmacology (1994-01-01)
D J Sanger, B Zivkovic
ABSTRACT
Alpidem in an imidazopyridine derivative which binds selectively to the omega 1 (BZ1) receptor subtype. It is active in some, but not all, behavioural tests sensitive to benzodiazepine anxiolytics and has clinical anti-anxiety effects. However, in a previous study, it was shown that alpidem did not substitute for chlordiazepoxide in rats trained to discriminate this benzodiazepine. The present experiments were carried out to investigate the discriminative stimulus properties of alpidem in greater detail. In the first experiment rats learned to discriminate a dose of 10 mg/kg alpidem from saline. Acquisition of the discrimination was long and performance unstable. Chlordiazepoxide, clorazepate and zolpidem substituted only partially for alpidem but the effects of the training dose of alpidem were blocked by 10 mg/kg flumazenil. The second experiment established stimulus control more rapidly to a dose of 30 mg/kg alpidem. Alpidem induced dose-related stimulus control, and dose-related and complete substitution for alpidem was produced by zolpidem, abecarnil, CL 218,872, triazolam and suriclone. Partial substitution occurred with chlordiazepoxide, clorazepate and pentobarbital. In most cases, high levels of substitution were produced only by doses which greatly reduced response rates even though the training dose of alpidem produced only modest decreases in rates. Ethanol, buspirone and bretazenil produced very little substitution for alpidem and both flumazenil and bretazenil antagonised the effects of alpidem. In two further experiments alpidem was found to substitute for the stimulus produced by zolpidem (2 mg/kg) but not for that produced by ethanol (1.5 g/kg).(ABSTRACT TRUNCATED AT 250 WORDS)