MilliporeSigma
A rapid method for detecting barbiturates in serum using EI-SIM.
International journal of legal medicine (1994-01-01)
T Kojima, T Taniguchi, M Yashiki, T Miyazaki, Y Iwasaki, T Mikami, M Ohtani
ABSTRACT
A simple and rapid method for analysis of barbiturates in serum has been developed. In order to extract and clean barbiturates in serum, a separation column packed with Extrelut and Florisil was used, and the eluate was directly analyzed by means of electron impact selected ion monitoring (EI-SIM). Selected ions used were base peak ions of 10 barbituartes, and the internal standard used was allobarbital or secobarbital. The calibration curves were linear over the range 0.5-5 ng. Extraction of replicate serum samples containing 20 micrograms/1.5 ml and 5 micrograms/1.5 ml resulted in a recovery of 87.2-105.2% and 81.6-104.6%, respectively, with the exception of phenobarbital, which was 151.9% and 172.1%, respectively. Secobarbital was also analyzed in the serum of 13 patients who had been given secobarbital intravenously. In 3 out of 10 cases, secobarbital levels greater than 1 micrograms/ml were detected more than 72 h after administration. This method seems to have possibilities for clinical use.