MilliporeSigma
  • Acute systemic toxicity and antihypertensive activity of a novel todralazine analog in rats.
Acute systemic toxicity and antihypertensive activity of a novel todralazine analog in rats.
Acta poloniae pharmaceutica (1993-01-01)
Z Jastrzebski, H Czyzewska-Szafran, B Goźlińska, M Wutkiewicz, A Jedrych, Z Fijałek, K Biniecki, P Aranjo, L Ciszewski, A P Mazurek
ABSTRACT
Acute intravenous toxicity and antihypertensive activity of KB1, a novel todralazine analog was investigated and compared with the effects of todralazine (Td) in normotensive Wistar-Kyoto (WKY) and spontaneously hypertensive (SHR) rats. LD50 values were 72 mg.kg-1 for KB1, and 255 mg.kg-1 for Td in WKY and 43 mg.kg-1 or KB1 in SHR. Therefore, the toxicity of KB1 was higher than that of Td and it increased in SHR. The antihypertensive activity of KB1 (ED20% 9.8 mg.kg-1) in WKY was about 9 times less potent in comparison with Td (ED20% 1.1 mg.kg-1). Blood pressure reducing activity of KB1 augmented apparently in SHR (ED20% 2.5 mg.kg-1) whereas Td had not such an effect (ED20% 1.0). Thus, the influence of Td on blood pressure was similar in normotensive and hypertensive animals. Our results indicate that KB1 is capable of reducing blood pressure preferentially in hypertension.