MilliporeSigma
  • Probing of coenzyme quinone binding site of mitochondrial NADH:CoQ reductase by fluorescence dynamics.
Probing of coenzyme quinone binding site of mitochondrial NADH:CoQ reductase by fluorescence dynamics.
Biochemistry (1994-08-16)
I Ahmed, G Krishnamoorthy
ABSTRACT
The coenzyme quinone (CoQ) binding region of mitochondrial NADH:CoQ reductase (complex-I) was investigated by the fluorescent probes erythrosine-5'-iodoacetamide (ER) and 3,3'-diethyloxadicarbocyanine iodide (DODCI). Both steady-state and time-resolved fluorescence was used in these experiments. Both probes competed for the binding site of 2,3-dimethoxy-5-methyl-6-decyl-1,4-benzoquinone (DB), an analogue of CoQ. The fluorescence lifetimes of the complex-I bound probes were approximately 600 ps and approximately 1.7 ns in the cases of ER and DODCI, respectively. Binding of the probes was not affected by the binding of the inhibitor rotenone. However, rotenone binding caused some changes in the lifetime of the bound probes. Reduction of the enzyme caused an increase in the level of binding of ER and a decrease in the level of binding of DODCI. The level of binding of cationic DODCI increased with the increase in pH, and in the case of anionic of ER the trend was reverse. Binding of Ca2+ to complex-I resulted in an increase in the level of binding of ER and a decrease in the level of binding of DODCI. Reaction with N,N'-dicyclohexylcarbodiimide (DCCD) resulted in alterations in the time-resolved fluorescence profiles of dye: complex-I system. All these results were interpreted as due to the presence of carboxyl group(s) with pKa approximately 6 in the probe/CoQ binding region. The rotational correlation time (tau r) of DODCI bound at the CoQ region was 2-3 ns.(ABSTRACT TRUNCATED AT 250 WORDS)