MilliporeSigma
  • Synthesis and bioactivity of new phosphorodithioates derived from N-substituted maleamic esters.
Synthesis and bioactivity of new phosphorodithioates derived from N-substituted maleamic esters.
Comparative biochemistry and physiology. C, Comparative pharmacology and toxicology (1993-01-01)
S A de Licastro, G Wallace, E Seccacini, P Gonzalez Audino, E Zerba
ABSTRACT
1. A new series of organophosphorus compounds derived from N-substituted maleamic esters by reaction with dimethyl and diethyl phosphorodithioic acid was synthesized. 2. Two isomers, with chemical structures analogous to malathion were obtained for each maleamic ester assayed. 3. They were characterized, and their toxicity against a resistant strain of Musca domestica (strain G), a susceptible one (strain RAC) and Triatoma infestans (vector of Chagas' disease) were measured. 4. The structural chemical modification introduced by replacing a carboxyester group for a N-substituted amido one, reverted the resistance obtained for malathion. 5. Replacement of methyl by ethyl dithiophosphoric acid did not introduce significative differences. 6. In the case of Triatoma infestans, the synthesized compounds were less active than malathion.
Sigma-Aldrich
Maleamic acid