MilliporeSigma
  • Correlation of the anticholinesterase activity of a series of organophosphates with their ability to compete with agonist binding to muscarinic receptors.
Correlation of the anticholinesterase activity of a series of organophosphates with their ability to compete with agonist binding to muscarinic receptors.
Toxicology and applied pharmacology (1993-10-01)
T R Ward, D J Ferris, H A Tilson, W R Mundy
ABSTRACT
Some compounds that inhibit acetylcholinesterase (AChE) activity compete directly with quinuclidinyl benzilate (QNB) binding, a muscarinic antagonist which binds to all subtypes equally, and with cis-methyldioxolane (CD), an agonist that binds with high affinity to the M2 subtype of muscarinic receptors. The relationship between inhibition of AChE activity and the capability to affect muscarinic receptors directly has not been systematically explored. The interaction of eight organophosphates with muscarinic receptors was compared to their ability to inhibit AChE activity in vitro in tissue homogenates from rat hippocampus and frontal cortex, two cholinergically enriched areas of the brain. Of the compounds tested only echothiophate competed for [3H]QNB binding and only at concentrations greater than 100 microM. The anticholinesterase compounds were also tested for their ability to compete with a muscarinic receptor agonist, [3H]CD, which binds with high affinity (approximate KD = 3.5 nM) to 10 and 3% of the muscarinic receptors in the frontal cortex and hippocampus, respectively. The anticholinesterase compounds inhibited high-affinity [3H]CD binding up to 80% and the effects were similar in both tissues. Echothiophate and DFP were potent inhibitors of [3H]CD binding, as were the active "oxon" forms of parathion, malathion, and disulfoton. The parent "thio" forms of these insecticides, however, were much less effective in competing for [3H]CD binding. A similar pattern of potency was observed for the inhibition of brain AChE activity. A strong correlation was found between the ability of a compound to inhibit AChE activity and the ability to compete with [3H]CD binding. These data suggest that the biological effects of cholinesterase-inhibiting compounds may be due to more than their ability to inhibit AChE.
Sigma-Aldrich
2-Methyl-1,3-dioxolane, 97%
Pricing and availability is not currently available.