MilliporeSigma
  • Suppression of lipopolysaccharide-induced fulminant hepatitis and tumor necrosis factor production by bisbenzylisoquinoline alkaloids in bacillus Calmette-Guerin-treated mice.
Suppression of lipopolysaccharide-induced fulminant hepatitis and tumor necrosis factor production by bisbenzylisoquinoline alkaloids in bacillus Calmette-Guerin-treated mice.
Biochemical pharmacology (1993-11-17)
Y Kondo, F Takano, H Hojo
ABSTRACT
The bisbenzylisoquinoline (BBI) alkaloids chondocurine, cycleanine, tetrandrine and berbamine were tested for their capacity to suppress hepatic injury and production of tumor necrosis factor (TNF) induced by lipopolysaccharide (LPS) in mice primed with bacillus Calmette-Guerin (BCG). When administered for three consecutive days before LPS injection, chondocurine, cycleanine and tetrandrine (10 mg/kg/day) strongly suppressed serum alanine aminotransferase (EC 2.6.1.1.) and aspartate aminotransferase (EC 2.6.1.2.); however, berbamine gave only slight protection. Chondocurine, cycleanine and tetrandrine but not berbamine significantly reduced the level of TNF which peaked 2 hr after LPS injection. This study shows that BBI alkaloids prevent BCG/LPS-induced hepatitis at least in part by suppressing TNF production.