MilliporeSigma
  • Preparation and application of a fluorogenic substrate for endo-beta-xylosidase.
Preparation and application of a fluorogenic substrate for endo-beta-xylosidase.
Journal of biochemical and biophysical methods (1989-08-01)
K Takagaki, A Kon, H Kawasaki, T Nakamura, M Endo
ABSTRACT
The present paper describes a fluorometric assay for galactosaminoglycan-degrading endo-beta-xylosidase, utilizing glycosaminoglycan chains bearing a 4-methylumbelliferyl group at the reducing terminus as a substrate. This fluorogenic substrate is synthesized by human skin fibroblasts cultured in the presence of a fluorogenic xyloside, 4-methylumbelliferyl-beta-D-xyloside. The assay is based on measurement of the fluorescence of 4-methylumbelliferone, enzymatically liberated from the synthetic substrate by endo-beta-xylosidase. We examined the applicability of the assay for analysis of endo-beta-xylosidase activity.