MilliporeSigma
  • Detection of free radicals in living mice after inhalation of DTBN by X-band ESR.
Detection of free radicals in living mice after inhalation of DTBN by X-band ESR.
Free radical biology & medicine (1993-03-01)
K Mäder, R Stösser, H H Borchert
ABSTRACT
A new application of X-band electron spin resonance (ESR) is demonstrated for in vivo investigations on living mice. It is the first report on in vivo detection, identification, and localization of a free radical after uptake in the body of a living mammal by inhalation. The volatile di-tert-butyl-nitroxyl (DTBN) has been used as a suitable spin probe. After inhalation of DTBN, a superimposed ESR spectrum consisting of a polar (g = 2.0057; a = 1.65 mT) and a nonpolar paramagnetic species (g = 2.0061; a = 1.51 mT) was detected in mice tails. Collected blood exhibits only the polar spectrum. The decay of signals after removing of the DTBN-soaked swab follows first-order kinetics with k = 0.08 min-1.