MilliporeSigma
  • The effect of p-chloroaniline on leucocytes of sheep peripheral blood under the migration-inhibition test conditions.
The effect of p-chloroaniline on leucocytes of sheep peripheral blood under the migration-inhibition test conditions.
Immunopharmacology and immunotoxicology (1995-08-01)
P Kacmár, J Pistl, I Mikula
ABSTRACT
Toxic and immunotoxic effects of p-chloroaniline--a metabolite of herbicide monolinuron--were investigated in peripheral blood leucocytic suspensions of five sheep using a migration-inhibition test. The toxic effect of p-chloroaniline was recorded at concentrations 1.0 to 0.1 mg.ml-1 and the immunotoxic one at concentrations 0.01-0.001 mg.l-1. The toxic effect was demonstrated by total inhibition of leucocyte migration. The immunotoxic effect, determined as mitogenic activation of leucocytes by phytohemagglutinin, concanavalin A and lipopolysaccharide, was detected at 10 to 100-fold lower concentrations of p-chloroaniline than those which resulted in toxic effects.