MilliporeSigma
  • Amine effect on phenylboronic acid complex with glucose under physiological pH in aqueous solution.
Amine effect on phenylboronic acid complex with glucose under physiological pH in aqueous solution.
Journal of biomaterials science. Polymer edition (1996-01-01)
D Shiino, A Kubo, Y Murata, Y Koyama, K Kataoka, A Kikuchi, Y Sakurai, T Okano
ABSTRACT
A new "intelligent' polymer system was developed utilizing the binding and exchange of phenylboronic acid (PBA) with polyols and/or glucose. In this improved system, an amine component was incorporated into the polymer chain along with PBA, to enhance binding between PBA and glucose under physiological conditions. The PBA-based polymer was formed by free-radical copolymerization of 3-methacrylamidophenylboronic acid (MAPB) with comonomers, N,N-dimethylaminopropylacrylamide (DMAPAA) and acrylamide (AAm) in the presence of N,N'-methylenebis(acrylamide) (Bis-AAm) as a cross-linker. The proportion of the amount of PBA groups complexed with glucose vs total amount of PBA groups was determined by the batch method. Compared to PBA copolymers synthesized without amine component, the proportion increased as a function of the amine content as well as the pH of the buffer. These results confirm that the interaction of neighboring amines (unprotonated) to PBA strengthens the binding with glucose, especially at pH 7.4 and above. This new PBA-amine copolymer is promising as a material useful for polyol separation, protection of polyols, and possibly, as an insulin delivery device.