MilliporeSigma
  • Composition of the Lactobacillus acidophilus complex isolated from vaginal flora.
Composition of the Lactobacillus acidophilus complex isolated from vaginal flora.
The new microbiologica (1996-04-01)
N Lachlak, E Ageron, O Zampatti, G Michel, P A Grimont
ABSTRACT
One hundred and fifty human vaginal samples containing a diversity of pathogens or nonpathogens (Gardnerella vaginalis, Streptococcus sp., Staphylococcus sp., Candida albicans. Mycoplasma sp.) were examined for their content in lactobacilli of the Lactobacillus acidophilus complex. Although all samples contained lactobacilli, strains of the L. acidophilus complex were present in only twenty-nine cases. Isolates were further characterized and compared with type strains or reference strains in an attempt to differentiate by phenotypic means the genospecies of the L. acidophilus complex. Data regarding specific activities of beta-galactosidase (beta-gal) and of phospho-beta-galactosidase (P-beta-gal) provided no specific information at the species level within the L. acidophilus complex. DNA-relatedness differentiates this genospecies. Most lactobacilli isolated from the vaginal flora of symptomatic women were genotypically close to L. gasseri CIP 102991T by the technique of DNA/DNA hybridization.