MilliporeSigma
  • Technetium-99m radiolabeled ouabagenin-cysteine conjugate: biological evaluation in animal models.
Technetium-99m radiolabeled ouabagenin-cysteine conjugate: biological evaluation in animal models.
Nuclear medicine and biology (1996-02-01)
M Chatterjee, S Ganguly, B R Sarkar, S Banerjee
ABSTRACT
Two ouabagenin-cysteine conjugates were synthesized by condensing 3-beta monochloroacetyl and 3-beta, 11-alpha dichloroacetyl ouabagenin with cysteine. The resulting ligands were radiolabeled with technetium-99m (99mTc) to furnish a single homogenous 99mTc chelate in each case with good stability. The animal experiments with these 99mTc-labeled conjugates established the superiority of guinea pig over rat and rabbit as an animal model, as previously observed for other tritiated or radioidinated cardiac glycosides or aglycones. In biodistribution experiments in guinea pig, these 99mTc chelates showed a favorable heart to liver (and other nontarget organ) uptake ratio, comparable to that of recently reported 125I-digoxigenin iodohistamine-3-oxime. The low heart to blood ratio in animal experiments with ouabagenin derivatives could be attributed to the absence of 3-beta sugar residues in these molecules, which is in agreement with the previous observation reported in connection with radioiodinated digoxin and digoxigenin derivatives.