MilliporeSigma
  • Application of arylosulfonic acids in analysis of antihypertensive drugs.
Application of arylosulfonic acids in analysis of antihypertensive drugs.
Acta poloniae pharmaceutica (1995-01-01)
M Gajewska, M Sokołowska-Muszyńska, J Iliaszenko, A Somogi
ABSTRACT
Suitability of four arylosulfonic acids and their sodium salts to form derivatives with hypotensive drugs were studied. New crystalline arylosulfonates of todralazine, hydralazine and dihydralazine were obtained. Physico-chemical properties of the obtained arylosulfonates were tested. Reagents mentioned above were also used in analysis of these drugs.