MilliporeSigma
  • Interactions of Ginkgo biloba extract (EGb 761), diazepam and ethyl beta-carboline-3-carboxylate on social behavior of the rat.
Interactions of Ginkgo biloba extract (EGb 761), diazepam and ethyl beta-carboline-3-carboxylate on social behavior of the rat.
Pharmacology, biochemistry, and behavior (1997-02-01)
R Chermat, D Brochet, F V DeFeudis, K Drieu
ABSTRACT
The social interaction test was used to examine the effects of an extract of Ginkgo biloba (EGb 761) and its possible interactions with diazepam and ethyl beta-carboline-3-carboxylate (beta-CCE). Pairs of naive (unfamiliar) male Wistar AF rats subjected to the same treatment were placed in a novel test arena that was brightly illuminated, and the duration (in s) of social contact was observed over a 10 min period. Single injections of EGb 761 (8-16 mg/kg, i.p.), given 30 min prior to testing, or repeated oral administration of the extract (48 or 96 mg/kg/day) for 8 days, significantly decreased social contact under conditions that did not influence locomotor activity. Injection of diazepam (1 mg/kg, i.p.), 30 min before testing, significantly increased social contact. Injection of diazepam to animals that had received repeated oral treatment with EGb 761 (96 mg/kg/ day) increased social interaction to an extent greater than observed with diazepam alone. Injection of beta-CCE (2-16 mg/kg, i.p.), 15 min before testing, significantly decreased social contact. When the animals were treated with EGb 761 (48 or 96 mg/kg/day, p.o. for 8 days) and beta-CCE (4 mg/kg), both of which decreased social interaction when administered alone, the resulting level of social contact was similar to that of control animals. Interactions with certain sites of central GABAA/ benzodiazepine/Cl- channel receptor complexes could be involved in mediating these effects of EGb 761, diazepam and beta-CCE.