MilliporeSigma
  • Primary targets for photoinactivation of vesicular stomatitis virus by AIPcS4 or Pc4 and red light.
Primary targets for photoinactivation of vesicular stomatitis virus by AIPcS4 or Pc4 and red light.
Photochemistry and photobiology (1997-03-01)
A C Moor, A E Wagenaars-van Gompel, A Brand, M A Dubbelman, J VanSteveninck
ABSTRACT
Phthalocyanines are useful sensitizers for the photodynamic sterilization of red blood cell concentrates. The mechanism of photoinactivation of lipid-enveloped viruses is not completely understood. Vesicular stomatitis virus (VSV) was used as a model virus to study the primary targets of photoinactivation by aluminum phthalocyanine tetrasulfonate (AIPcS4) or silicon phthalocyanine HOSiPcOSi(CH3)2(CH2)3N(CH3)2 (Pc4) and red light. Inactivation conditions for VSV in buffer were determined using an end point dilution assay, and viral RNA synthesis in host cells was measured to determine the loss of infectivity in a direct way. The very rapid decrease in the viral RNA synthesis after photodynamic treatment was correlated with respect to different potential primary targets that are involved in different steps of the viral replication cycle. Damage to the viral proteins, induced by treatment with AIPcS4 or Pc4 and analyzed by gel electrophoresis, could not account for the observed loss of infectivity. Binding of VSV to host cells was only slightly impaired after photodynamic treatment with both sensitizers and could therefore not be responsible for the rapid decrease in viral RNA synthesis in cells. A very strong inhibition of viral RNA polymerase activity after treatment with AIPcS4 and red light was detectable using an in vitro assay. This decrease correlated well with the loss of infectivity, indicating that either the RNA or the viral RNA polymerase is the primary target for photoinactivation of VSV with AIPcS4. Treatment with Pc4 did not cause inhibition of viral RNA polymerase activity to an extent that could account for the observed very rapid loss of infectivity. It was therefore concluded that neither the viral proteins nor the binding to the host cells nor the RNA or RNA polymerase are the primary targets for photoinactivation of VSV by Pc4.