MilliporeSigma
  • Design of potent dynorphin A-(1-9) analogues devoid of supraspinal motor effects in mice.
Design of potent dynorphin A-(1-9) analogues devoid of supraspinal motor effects in mice.
Canadian journal of physiology and pharmacology (1997-01-01)
H T Le, R Michelot, M Dumont, V K Shukla, M Mayer, K P Nguyen, H Ruan, S Lemaire
ABSTRACT
Four analogues of dynorphin (Dyn) A-(1-9) incorporating D-Leu in position 8 alone or in combination with the nonhydrolysable psi [CS-NH] thiopeptide bond surrogate between positions 6 and 7 were tested in vitro for their ability to compete with the binding of selective kappa, mu, and delta opioid ligands, using membrane preparations of guinea pig cerebellum (kappa) and rat brain (mu and delta), for their ability to block the electrically induced contractions of the guinea pig ileum, and for their in vivo antinociceptive (writhing test) and motor (motor dysfunction assay) activities in mice. [D-Leu8]Dyn A-(1-9) displayed an affinity and a selectivity for the kappa opioid receptor that were comparable with those of Dyn A-(1-9). The potencies of [D-Leu8]Dyn A-(1-9) in the guinea pig ileum, writhing, and motor dysfunction assays were markedly enhanced (8-12 fold) compared with those of Dyn A-(1-9). [6 psi 7(CS-NH),D-Leu8]Dyn A-(1-9), [Lys6, 6 psi 7(CS-NH),D-Leu8] Dyn A-(1-9), and [Leu6, 6 psi 7(CS-NH), D-Leu8]Dyn A-(1-9) were somewhat less potent than [D-Leu8]Dyn A-(1-9) in all opioid assays. However, the thiopeptides were more potent analgesics than Dyn A-(1-9)(ED50 of 29.5, 23.9, and 15.5 nmol/mouse, respectively, compared with 90.7 nmol/mouse for Dyn A-(1-9)) and caused little or no motor impairment at analgesic doses.