MilliporeSigma
  • Abnormal ventricular repolarization mimicking myocardial infarction after heterocyclic antidepressant overdose.
Abnormal ventricular repolarization mimicking myocardial infarction after heterocyclic antidepressant overdose.
The American journal of cardiology (1997-01-15)
R Bolognesi, D Tsialtas, P Vasini, M Conti, C Manca
ABSTRACT
In 2 young adult women who experienced acute heterocyclic antidepressant intoxication, we found a quite unusual electrocardiographic pattern characterized by abnormal ST-tract elevation in the right precordial leads associated with a marked QRS widening (right bundle branch block and left anterior fascicular block type). Because serum electrolyte imbalance and acute myocardial ischemic events were excluded, the mechanism by which antidepressant overdose may produce such elevation of the ST tract remains unclear.