MilliporeSigma
  • Determination of the enantiomeric composition of ibuprofen in human plasma by high-performance liquid chromatography.
Determination of the enantiomeric composition of ibuprofen in human plasma by high-performance liquid chromatography.
Journal of chromatography (1987-03-20)
A Avgerinos, A J Hutt
ABSTRACT
A normal-phase high-performance liquid chromatographic method, using a hexane-ethyl acetate solvent system, for the determination of the enantiomeric composition of ibuprofen in human plasma is described. The method is based on the resolution of the diastereoisomeric amides formed on reaction of the ibuprofen enantiomers with S-1-(naphthen-1-yl)ethylamine using p-chlorophenoxy-acetic acid as internal standard. The application of the method for the determination of the enantiomeric composition of ibuprofen in human plasma following the repeated oral administration of the drug to two volunteers is reported. The plasma concentrations of the S-(+) enantiomer were always greater than that of the R-(-), the ratio of the areas under the enantiomer plasma concentration-time curves (S/R) being 1.8 and 1.6.