MilliporeSigma
  • Terpenoid biosynthesis from 1-deoxy-D-xylulose in higher plants by intramolecular skeletal rearrangement.
Terpenoid biosynthesis from 1-deoxy-D-xylulose in higher plants by intramolecular skeletal rearrangement.
Proceedings of the National Academy of Sciences of the United States of America (1997-10-06)
D Arigoni, S Sagner, C Latzel, W Eisenreich, A Bacher, M H Zenk
ABSTRACT
The incorporation of [1-13C]- and [2,3,4,5-13C4]1-deoxy-D-xylulose into beta-carotene, lutein, phytol, and sitosterol in a cell culture of Catharanthus roseus was analyzed by NMR spectroscopy. The labeling patterns of the isoprene precursors, isopentenyl pyrophosphate and dimethylallyl pyrophosphate, were obtained from the terpenes by a retrobiosynthetic approach. 13C Enrichment and 13C13C coupling patterns showed conclusively that 1-deoxy-D-xylulose and not mevalonate is the predominant isoprenoid precursor of phytol, beta-carotene, and lutein. Label from 1-deoxyxylulose was also diverted to phytosterols to a minor extent (6% relative to carotene and phytol formation). The data demonstrate that the formation of isopentenyl pyrophosphate from pentulose occurs strictly by an intramolecular rearrangement process.