MilliporeSigma
[Antihypoxants: their status and outlook].
Eksperimental'naia i klinicheskaia farmakologiia (1998-10-23)
V D Luk'ianchuk, L V Savchenkova
ABSTRACT
The survey deals with the up-to-date information in the literature on the problem of pharmacological correction of pathological changes found in the organism in the hypoxic syndrome. The principal approaches to pharmacotherapy of this pathological condition are pointed out. Information on the up-to-date and effective antihypoxia agents applied in medical practice and those in the stage of development is given.
Sigma-Aldrich
2-Imino-4-thiobiuret, 99%