MilliporeSigma
  • [The vasoconstrictor effect of cyclosporine on the pulmonary vein is mediated by its vehicle: the cremophor].
[The vasoconstrictor effect of cyclosporine on the pulmonary vein is mediated by its vehicle: the cremophor].
Annales de chirurgie (1998-12-10)
P Mathieu, P Demers, S Elkouri, R Cartier
ABSTRACT
Cyclosporin A (CyA) induced vasoconstriction and impaired relaxation to agonists has been related in some models to its vehicle: the cremophor (CRE). There is no data concerning the effect of CyA and its vehicle CRE on the pulmonary veins. Therefore, the present study was designed to characterize the effect of CyA and CRE on isolated pulmonary veins. Third-order canine pulmonary veins (n = 6) were suspended in organ chambers for measurement of isometric force. Segments were exposed to cumulative doses (10(-9) to 10(-4) M) of CyA in its vehicle CRE, and to CRE alone. CyA induced an ehdothelium-independent vaasoconstriction similar to CRE, maximal constriction (Emax) were: 1.8 +/- 0.3 g versus 2.1 +/- 0.4 g respectively (p = NS). This vasoconstriction observed with CRE was not affected by indomethacine (a cyclooxygenase inhibitor), and by pinane thromboxane (a thromboxane antagonist). However, the vasoconstriction was decreased by diltiazem (10(-5) M), a calcium channel blocker, Emax were: 2.1 +/- 0.4 g and 0.8 +/- 0.04 g respectively for CRE and CRE with diltiazem (p < 0.05). CyA vehicle CRE induces an endothelium-independent vasoconstriction in isolated third order pulmonary veins in the dog. This vasoconstriction is not related to a cyclooxygenase product, but is partially mediated by a calcium channel activation in vascular smooth muscle.