MilliporeSigma
  • The cholesterol metabolite cholest-4-en-3-one and its 3-oxo derivatives suppress body weight gain, body fat accumulation and serum lipid concentration in mice.
The cholesterol metabolite cholest-4-en-3-one and its 3-oxo derivatives suppress body weight gain, body fat accumulation and serum lipid concentration in mice.
Bioorganic & medicinal chemistry letters (1999-01-05)
K Suzuki, T Shimizu, T Nakata
ABSTRACT
Based on the findings that cholest-4-en-3-one, an intestinal metabolite of cholesterol, has an anti-obesity effect on animals, the structure-effect relationship of its 3-oxo derivatives and related compounds were investigated. Cholesten-3-ones, which possesses an enone structure with a carbonyl group at C3, markedly inhibit body weight gain and body fat accumulation, as well as the levels of serum triglyceride and cholesterol in animals without any clinical abnormalities.
Sigma-Aldrich
(+)-4-Cholesten-3-one, 98%