MilliporeSigma
  • Selective regulation of IP3-receptor-mediated Ca2+ signaling and apoptosis by the BH4 domain of Bcl-2 versus Bcl-Xl.
Selective regulation of IP3-receptor-mediated Ca2+ signaling and apoptosis by the BH4 domain of Bcl-2 versus Bcl-Xl.
Cell death and differentiation (2011-08-06)
G Monaco, E Decrock, H Akl, R Ponsaerts, T Vervliet, T Luyten, M De Maeyer, L Missiaen, C W Distelhorst, H De Smedt, J B Parys, L Leybaert, G Bultynck
ABSTRACT
Antiapoptotic B-cell lymphoma 2 (Bcl-2) targets the inositol 1,4,5-trisphosphate receptor (IP(3)R) via its BH4 domain, thereby suppressing IP(3)R Ca(2+)-flux properties and protecting against Ca(2+)-dependent apoptosis. Here, we directly compared IP(3)R inhibition by BH4-Bcl-2 and BH4-Bcl-Xl. In contrast to BH4-Bcl-2, BH4-Bcl-Xl neither bound the modulatory domain of IP(3)R nor inhibited IP(3)-induced Ca(2+) release (IICR) in permeabilized and intact cells. We identified a critical residue in BH4-Bcl-2 (Lys17) not conserved in BH4-Bcl-Xl (Asp11). Changing Lys17 into Asp in BH4-Bcl-2 completely abolished its IP(3)R-binding and -inhibitory properties, whereas changing Asp11 into Lys in BH4-Bcl-Xl induced IP(3)R binding and inhibition. This difference in IP(3)R regulation between BH4-Bcl-2 and BH4-Bcl-Xl controls their antiapoptotic action. Although both BH4-Bcl-2 and BH4-Bcl-Xl had antiapoptotic activity, BH4-Bcl-2 was more potent than BH4-Bcl-Xl. The effect of BH4-Bcl-2, but not of BH4-Bcl-Xl, depended on its binding to IP(3)Rs. In agreement with the IP(3)R-binding properties, the antiapoptotic activity of BH4-Bcl-2 and BH4-Bcl-Xl was modulated by the Lys/Asp substitutions. Changing Lys17 into Asp in full-length Bcl-2 significantly decreased its binding to the IP(3)R, its ability to inhibit IICR and its protection against apoptotic stimuli. A single amino-acid difference between BH4-Bcl-2 and BH4-Bcl-Xl therefore underlies differential regulation of IP(3)Rs and Ca(2+)-driven apoptosis by these functional domains. Mutating this residue affects the function of Bcl-2 in Ca(2+) signaling and apoptosis.