MilliporeSigma
  • Nitric oxide elicits functional MMP-13 protein-tyrosine nitration during wound repair.
Nitric oxide elicits functional MMP-13 protein-tyrosine nitration during wound repair.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2008-05-23)
Tania R Lizarbe, Concepción García-Rama, Carlos Tarín, Marta Saura, Enrique Calvo, Juan Antonio López, Carlos López-Otín, Alicia R Folgueras, Santiago Lamas, Carlos Zaragoza
ABSTRACT
Nitric oxide (NO) plays a critical role in wound healing, in part by promoting angiogenesis. However, the precise repair pathways affected by NO are not well defined. We now show that NO regulates matrix metalloproteinase-13 (MMP-13) release during wound repair. We find that normally MMP-13 is kept inside endothelial cells by an association with caveolin-1. However, nitration of MMP-13 on tyrosine residue Y338 causes it to dissociate from caveolin-1 and be released from endothelial cells. We next explored the functional significance of MMP-13 nitration in vivo. Skin injury increases nitration of MMP-13 in mice. Skin wounds in inducible nitric oxide synthase knockout mice release less MMP-13 and heal more slowly than skin wounds in wild-type mice. Conversely, skin wounds in caveolin-1 knockout mice have increased NO production, increased MMP-13 nitration, and accelerated wound healing. Collectively, our data reveal a new pathway through which NO modulates wound repair: nitration of MMP-13 promotes its release from endothelial cells, where it accelerates angiogenesis and wound healing.