MilliporeSigma
  • Hydroxyl radical production and human DNA damage induced by ferric nitrilotriacetate and hydrogen peroxide.
Hydroxyl radical production and human DNA damage induced by ferric nitrilotriacetate and hydrogen peroxide.
Cancer research (1987-12-15)
S Inoue, S Kawanishi
ABSTRACT
Reactivities of Fe3+ chelates of aminopolycarboxylic acids with DNA were investigated by the DNA-sequencing technique using 32P 5'-end-labeled DNA fragments obtained from the human c-Ha-ras-1 protooncogene, and the reaction mechanism was studied by electron spin resonance spectroscopy. Ferric nitrilotriacetate (Fe3+-NTA) plus hydrogen peroxide caused strong DNA cleavage in the presence of albumin. No or little DNA cleavage was observed with ferric chloride or Fe3+ chelates of other aminopolycarboxylic acids tested in the presence of hydrogen peroxide. The DNA cleavage by Fe3+-NTA plus hydrogen peroxide without piperidine treatment occurred at positions of every nucleotide although a specific cleavage was observed, whereas cleavages at the positions of guanine and thymine increased predominantly with piperidine treatment. Electron spin resonance studies using free radical traps demonstrated that of Fe3+ chelates of aminopolycarboxylic acids, Fe3+-NTA was the most effective catalyst in hydrogen peroxide-derived production of hydroxyl radicals under our conditions. The results suggest that Fe3+-NTA catalyzes the decomposition of hydrogen peroxide to produce hydroxyl radicals, which subsequently cause the strong base alterations of guanine and thymine, and deoxyribose-phosphate backbone breakages. The possibility that the Fe3+-NTA-induced DNA damage is the initiation and/or promotion of carcinogenesis by Fe3+-NTA is discussed.