MilliporeSigma
  • A new ionic liquid dimethyldinonylammonium bromide as a flow modifier for the simultaneous determination of eight carboxylates by capillary electrophoresis.
A new ionic liquid dimethyldinonylammonium bromide as a flow modifier for the simultaneous determination of eight carboxylates by capillary electrophoresis.
Journal of chromatography. A (2005-11-09)
Pirkko-Leena Laamanen, Sara Busi, Manu Lahtinen, Rose Matilainen
ABSTRACT
Two new methods of capillary zone electrophoresis based on aqueous phosphate running buffers with UV spectrophotometric detection were developed and optimized for the determination of eight carboxylates as copper complexes. Metalcomplexes are negatively charged, so measurements were made as anion analyses with flow reversal in the capillary. Two flow modifiers were used: a common tetradecyltrimethylammonium bromide (TTAB) and a new ionic liquid dimethyldinonylammonium bromide (DMDNAB). The methods were compared to each other. Better separation was achieved with DMDNAB as the flow modifier. Method development was done using a fused silica capillary (61 cm x 50 microm i.d.). Optimization was done using 95 mmol L(-1) phosphate buffer with TTAB or DMDNAB in the concentration 0.5 mmol L(-1) at pH 7.1. A -20 kV voltage and direct UV detection at 254 nm was used in measurements. In both CE methods all the peaks in the electropherograms were properly separated, the calibration plots gave good correlation coefficients and all eight carboxylates were detected in less than 7.5 min. The two methods were tested with natural water samples and a paper mill sample, and proved to be feasible.