MilliporeSigma
  • A unique glycosyltransferase involved in the initial assembly of Moraxella catarrhalis lipooligosaccharides.
A unique glycosyltransferase involved in the initial assembly of Moraxella catarrhalis lipooligosaccharides.
Glycobiology (2008-03-14)
Johanna M Schwingel, Frank St Michael, Andrew D Cox, Hussein Masoud, James C Richards, Anthony A Campagnari
ABSTRACT
Moraxella catarrhalis express three predominant forms of lipooligosaccharide (LOS) molecules on the bacterial surface. These major glycolipids contain specific carbohydrate epitopes that distinguish each glycoform into serotype A, B, or C LOS. All three serotypes, however, share a common glucose containing inner-core structure, consisting of an alpha-glucose attached to 2-keto-3-deoxyoctulosonic acid (KDO), which is unique among Gram-negative bacteria. Many of the LOS glycosyltransferase genes (lgt) responsible for assembly of the extended M. catarrhalis LOS structure have been identified. In this report, we now describe the identification and characterization of Lgt6, a unique glycosyltransferase that is responsible for the addition of the first glucose to the inner core thus initiating the assembly of full length LOS. Isogenic mutants defective in the expression of lgt6 were constructed in all three M. catarrhalis LOS serotypes and the resulting LOS glycoforms consisted of KDO(2)-lipid A-OH as analyzed by urea sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) and mass spectrometry. In addition, the expression of lgt6 in trans in a heptose-deficient Neisseria meningitidis NMB gmhX mutant resulted in the addition of a hexose to the LOS of this strain. These studies demonstrate that Lgt6 functions as an alpha-(1-5)-glucosyltransferase in M. catarrhalis adding the primary glucose to the KDO(2)-lipid A-OH in LOS biosynthesis. The function of Lgt6 is required for the completion of both the major and minor oligosaccharide chains in M. catarrhalis.