MilliporeSigma
The presence of free D-serine, D-alanine and D-proline in human plasma.
Experientia (1992-10-15)
Y Nagata, R Masui, T Akino
ABSTRACT
Twelve neutral free amino acids, i.e. serine, threonine, glutamine, asparagine, alanine, proline, methionine, tyrosine, valine, leucine, isoleucine and phenylalanine, were surveyed for the presence of D-enantiomers in plasma samples from patients with renal diseases and from normal subjects. D-serine, D-alanine and D-proline were found in the patient's plasma. The highest concentrations (D/L ratio) of D-serine, D-alanine and D-proline were 0.2362, 0.2087 and 0.0986, respectively. The sum of the contents of the three D-amino acid was shown to be present in the plasma proteins.
Sigma-Aldrich
D-Alanine, ≥98% (HPLC)