MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Long-term validity of biological markers of psychopathy and criminal recidivism: follow-up 6-8 years after forensic psychiatric investigation.
Long-term validity of biological markers of psychopathy and criminal recidivism: follow-up 6-8 years after forensic psychiatric investigation.
Psychiatry research (2003-12-17)
E Gunilla Stalenheim
ABSTRACT
This study is a follow-up investigation of a forensic psychiatric sub-population 6-8 years after forensic psychiatric evaluation. The aim was to examine the long-term validity of biological markers of psychopathy and antisocial behavior over time. Data on criminal records were obtained at follow-up from the National Council for Crime Prevention. Basic data included findings of psychiatric and psychological assessments, as well as values for serum triiodothyronine (T3) and free thyroxin (FT4), and platelet monoamine oxidase (MAO) activity, all obtained during the forensic psychiatric examination. Criminal recidivists at follow-up had higher serum T3 levels than non-recidivists, and much higher values than normal controls, while their levels of free T4 were lower. The T3 levels in criminal recidivists correlated to psychopathy- and aggression-related personality traits as measured by the Karolinska Scale of Personality. In violent recidivists, a remarkably high correlation was noted between T3 levels and Irritability and Detachment, traits that have previously been linked to the dopaminergic system. Stepwise multiple regression analyses confirmed the relationships of T3 levels and platelet MAO activity with personality traits in criminal recidivists. The predictive validity of biological markers of psychopathy, T3 and platelet MAO, measured during forensic psychiatric investigation, is stable over time. The results indicate chronic alterations of the hypothalamic-pituitary-thyroid axis in this group of subjects.
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine, ≥95% (HPLC), powder
Supelco
3,3′,5-Triiodo-L-thyronine (T3) solution, 100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®