MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • The prostate 25-hydroxyvitamin D-1 alpha-hydroxylase is not influenced by parathyroid hormone and calcium: implications for prostate cancer chemoprevention by vitamin D.
The prostate 25-hydroxyvitamin D-1 alpha-hydroxylase is not influenced by parathyroid hormone and calcium: implications for prostate cancer chemoprevention by vitamin D.
Carcinogenesis (2004-01-20)
Michael V Young, Gary G Schwartz, Lilin Wang, Daniel P Jamieson, Lyman W Whitlatch, John N Flanagan, Bal L Lokeshwar, Michael F Holick, Tai C Chen
ABSTRACT
The hormonal form of vitamin D, 1 alpha,25-dihydroxyvitamin D [1 alpha,25(OH)(2)D] promotes the differentiation and inhibits the proliferation, invasiveness and metastasis of prostate cells. However, 1 alpha,25(OH)(2)D is not suitable as a chemopreventive agent because its administration can cause hypercalcemia. Serum levels of 1 alpha,25(OH)(2)D are tightly regulated by the renal enzyme, 25-hydroxyvitamin D-1 alpha-hydroxylase (1 alpha-OHase), which synthesizes 1 alpha,25(OH)(2)D from the prohormone, 25-hydroxyvitamin D [25(OH)D]. Normal prostate epithelial cells in primary culture, as well as several prostate cancer cell lines, also express 1 alpha-OHase and synthesize the hormone intracellularly. We now investigated the regulation of the prostate 1 alpha-OHase by the three most important regulators of the renal 1 alpha-OHase: calcium, 1 alpha,25(OH)(2)D and parathyroid hormone (PTH). The 1 alpha-OHase activity in the primary cultures of prostate epithelial cells was inhibited by 1 alpha,25(OH)(2)D(3) at 10 and 100 nM, whereas PTH at 10 and 100 nM had no significant effect. Calcium at 1.2 and 2.4 mM had no significant effect on the enzyme activity in the PZ-HPV-7 cell line, a prostate epithelial cell line derived from normal prostate tissue. Conversely, 1.2 or 2.4 mM calcium markedly inhibited 1 alpha-OHase activity in a human kidney cell line used as a positive control. Furthermore, PTH at 100 nM and calcium at 1.2 and 2.4 mM had no effect on the 1 alpha-OHase gene promoter activity in prostate cells, whereas the promoter activity was inhibited 48 +/- 5% by 100 nM 1 alpha,25(OH)(2)D(3). Our findings suggest that, unlike the renal enzyme, the prostate 1 alpha-OHase appears to be largely unregulated by serum levels of PTH and calcium. These findings support the hypothesis that vitamin D or 25(OH)D may be useful as chemopreventive agents for prostate cancer because their administration should cause an increased synthesis of 1 alpha,25(OH)(2)D within prostate cells.
Product Description
Ergocalciferol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ergocalciferol, ≥98.0% (sum of enantiomers, HPLC)
Supelco
Ergocalciferol (Vitamin D2), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ergocalciferol, 40,000,000 USP units/g