MilliporeSigma
  • Beneficial effect of liver transplantation on bone mineral density in small infants with cholestasis.
Beneficial effect of liver transplantation on bone mineral density in small infants with cholestasis.
Transplantation proceedings (2004-07-15)
J Pawłowska, H Matusik, P Socha, H Ismail, J Ryzko, E Karczmarewicz, I Jankowska, M Teisseyre, R Lorenc
ABSTRACT
Reports of bone mineral density in children after liver transplantation are few. Eleven cholestatic children were analyzed before and 6 months after liver transplantation. No changes in serum levels of calcium, alkaline phosphates, or 25OHD were observed before versus after LTx. The serum levels of phosphorus and 1-25(OH)2D3 as well as total bone mass density and Cole index were significantly increased after liver transplantation.
Sigma-Aldrich
Phosphorus, red, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus, red, ≥97.0%