MilliporeSigma
  • Product distribution in amylase-catalyzed hydrolysis of amylose. Comparison of experimental results with theoretical predictions.
Product distribution in amylase-catalyzed hydrolysis of amylose. Comparison of experimental results with theoretical predictions.
Journal of biochemistry (1980-04-01)
H Kondo, H Nakatani, R Matsuno, K Hiromi
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)