MilliporeSigma
  • High-purity 4-nitrophenol: purification, characterization, and specifications for use as a spectrophotometric reference material.
High-purity 4-nitrophenol: purification, characterization, and specifications for use as a spectrophotometric reference material.
Clinical chemistry (1980-05-01)
G N Bowers, R B McComb, R G Christensen, R Schaffer
ABSTRACT
We describe specifications for high-purity 4-nitrophenol, which is suitable for spectrophotometric standardization. Such a reference material is needed in clinical enzymology to establish the proper molar absorptivity of 4-nitrophenol under final reaction conditions, particularly for measuring alkaline phosphatase activity in human serum. Some lots of 4-nitrophenol available commercially met these specifications, but several did not. The latter can be purified to meet our specifications by recrystallization or sublimation. The molar absorptivity of 4-nitrophenol (35 mumol/L) IN 10 mmol/L NaOH at 25 degrees C at 401 nm is 18380 +/- 90 L.mol-1.cm-1.
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Supelco
4-Nitrophenol, PESTANAL®, analytical standard