MilliporeSigma
[The relationship between serum leukotriene B4 and smoking].
Nihon eiseigaku zasshi. Japanese journal of hygiene (1993-08-01)
Y Yanagisawa, T Nagai
ABSTRACT
Leukotriene B4 (LTB4), an arachidonic acid metabolite released by neutrophils, is involved in the regulation of the host immune response to antigenic stimulation. Furthermore, LTB4 affects the chemokinesis, aggregation, and enzyme release of neutrophils and stimulates activity of cytotoxic T cells, natural killer cells and suppressor T cells. On the other hand, smoking typically results in inflammatory stimulation of the lung, and long-term smoking can cause chronic stimulation. In this paper, we report the measurement of LTB4, IgG, IgA, IgM, IgD and IgE in sera of cigarette smokers who did not have an allergic reaction (group 1), non-smokers who also had no history of allergy (group 2) and non-smokers who had delicate allergic conditions (group 3). The mean LTB4 concentration in serum of group 1 was nearly 3.3-fold greater than in group 2 and 2-fold greater than in group 3, being 430 +/- 55 (Mean +/- S.D.), 130 +/- 22 and 220 +/- 14 pg/ml, respectively. Concentrations of IgG, IgA, IgM, IgD and IgE of all volunteers were in the normal healthy range. The mean concentration of IgE in the serum of group 1 did not change significantly within one day. It was 35 +/- 4 IU/ml in the morning, 34 +/- 4 IU/ml in the afternoon, 31 +/- 2 IU/ml in the evening and 32 +/- 4 IU/ml the next morning. But in group 3 the IgE concentration changed significantly within one day, being < 25 IU/ml in the morning, 97 +/- 7 IU/ml in the afternoon, 97 +/- 7 IU/ml in the evening and 25 +/- 14 IU/ml the next morning.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
Leukotriene B4, ≥97%