MilliporeSigma
  • Improvement of accuracy in flow immunosensor system by introduction of poly-2-[3-(methacryloylamino) propylammonio] ethyl 3-aminopropyl phosphate.
Improvement of accuracy in flow immunosensor system by introduction of poly-2-[3-(methacryloylamino) propylammonio] ethyl 3-aminopropyl phosphate.
Journal of Sensors
Fuchiwaki Y, et al.