MilliporeSigma
  • Effect of decompressive craniectomy on aquaporin-4 expression after lateral fluid percussion injury in rats.
Effect of decompressive craniectomy on aquaporin-4 expression after lateral fluid percussion injury in rats.
Journal of neurotrauma (2010-11-19)
Satoshi Tomura, Hiroshi Nawashiro, Naoki Otani, Yoichi Uozumi, Terushige Toyooka, Atsushi Ohsumi, Katsuji Shima
ABSTRACT
Decompressive craniectomy is one therapeutic option for severe traumatic brain injury (TBI), and it has long been used for the treatment of patients with malignant post-traumatic brain edema. A lack of definitive evidence, however, prevents physicians from drawing any conclusions about the effects of decompressive craniectomy for the treatment of TBI. Therefore, the aim of the present study was to investigate the influence of decompressive craniectomy on post-traumatic brain edema formation. The aquaporin-4 (AQP4) water channel is predominantly expressed in astrocytes, and it plays an important role in the regulation of brain water homeostasis. In the present study, we investigated the time course of AQP4 expression and the water content of traumatized cortex following decompressive craniectomy after TBI. Adult male Sprague-Dawley rats (300-400 g) were subjected to lateral fluid percussion injury using the Dragonfly device. The effect of decompressive craniectomy was studied in traumatized rats without craniectomy (closed skull, DC-), and in rats craniectomized immediately after trauma (DC+). AQP4 expression was investigated with a Western blot analysis and immunohistochemistry. Brain edema was measured using the wet weight/dry weight method. At 48 h after TBI, AQP4 expression of the DC- group was significantly increased compared with the DC+ group (p < 0.01). In addition, the cortical water content of the DC- group was significantly increased compared to the DC+ group at the same time point (p < 0.05). The present results suggest that decompressive craniectomy may affect AQP4 expression and reduce brain edema formation after TBI.