MilliporeSigma
  • A core transcriptional network composed of Pax2/8, Gata3 and Lim1 regulates key players of pro/mesonephros morphogenesis.
A core transcriptional network composed of Pax2/8, Gata3 and Lim1 regulates key players of pro/mesonephros morphogenesis.
Developmental biology (2013-08-08)
Sami Kamel Boualia, Yaned Gaitan, Mathieu Tremblay, Richa Sharma, Julie Cardin, Artur Kania, Maxime Bouchard
ABSTRACT
Translating the developmental program encoded in the genome into cellular and morphogenetic functions requires the deployment of elaborate gene regulatory networks (GRNs). GRNs are especially crucial at the onset of organ development where a few regulatory signals establish the different programs required for tissue organization. In the renal system primordium (the pro/mesonephros), important regulators have been identified but their hierarchical and regulatory organization is still elusive. Here, we have performed a detailed analysis of the GRN underlying mouse pro/mesonephros development. We find that a core regulatory subcircuit composed of Pax2/8, Gata3 and Lim1 turns on a deeper layer of transcriptional regulators while activating effector genes responsible for cell signaling and tissue organization. Among the genes directly affected by the core components are the key developmental molecules Nephronectin (Npnt) and Plac8. Hence, the pro/mesonephros GRN links together several essential genes regulating tissue morphogenesis. This renal GRN sheds new light on the disease group Congenital Anomalies of the Kidney and Urinary Tract (CAKUT) in that gene mutations are expected to generate different phenotypic outcomes as a consequence of regulatory network deficiencies rather than threshold effects from single genes.