MilliporeSigma
  • Specific entrapment of methanol and dimethyl sulphoxide (DMSO) by a simple host compound (triphenylmethanol). Crystal structures of the Ph3COH? MeOH (1: 1) and Ph3COH? DMSO (2: 1) clathrate inclusion complexes.
Specific entrapment of methanol and dimethyl sulphoxide (DMSO) by a simple host compound (triphenylmethanol). Crystal structures of the Ph3COH? MeOH (1: 1) and Ph3COH? DMSO (2: 1) clathrate inclusion complexes.
Journal of the Chemical Society. Chemical Communications (1989)
Weber E, et al.