MilliporeSigma
  • The CREB coactivator TORC2 functions as a calcium- and cAMP-sensitive coincidence detector.
The CREB coactivator TORC2 functions as a calcium- and cAMP-sensitive coincidence detector.
Cell (2004-09-30)
Robert A Screaton, Michael D Conkright, Yoshiko Katoh, Jennifer L Best, Gianluca Canettieri, Shawn Jeffries, Ernesto Guzman, Sherry Niessen, John R Yates, Hiroshi Takemori, Mitsuhiro Okamoto, Marc Montminy
ABSTRACT
Elevations in circulating glucose and gut hormones during feeding promote pancreatic islet cell viability in part via the calcium- and cAMP-dependent activation of the transcription factor CREB. Here, we describe a signaling module that mediates the synergistic effects of these pathways on cellular gene expression by stimulating the dephosphorylation and nuclear entry of TORC2, a CREB coactivator. This module consists of the calcium-regulated phosphatase calcineurin and the Ser/Thr kinase SIK2, both of which associate with TORC2. Under resting conditions, TORC2 is sequestered in the cytoplasm via a phosphorylation-dependent interaction with 14-3-3 proteins. Triggering of the calcium and cAMP second messenger pathways by glucose and gut hormones disrupts TORC2:14-3-3 complexes via complementary effects on TORC2 dephosphorylation; calcium influx increases calcineurin activity, whereas cAMP inhibits SIK2 kinase activity. Our results illustrate how a phosphatase/kinase module connects two signaling pathways in response to nutrient and hormonal cues.