MilliporeSigma
  • Involvement of SIK2/TORC2 signaling cascade in the regulation of insulin-induced PGC-1alpha and UCP-1 gene expression in brown adipocytes.
Involvement of SIK2/TORC2 signaling cascade in the regulation of insulin-induced PGC-1alpha and UCP-1 gene expression in brown adipocytes.
American journal of physiology. Endocrinology and metabolism (2009-04-09)
Masaaki Muraoka, Aiko Fukushima, Say Viengchareun, Marc Lombès, Fukuko Kishi, Akira Miyauchi, Mariko Kanematsu, Junko Doi, Junko Kajimura, Ryo Nakai, Tatsuya Uebi, Mitsuhiro Okamoto, Hiroshi Takemori
ABSTRACT
Salt-inducible kinase 2 (SIK2) is expressed abundantly in adipose tissues and represses cAMP-response element-binding protein (CREB)-mediated gene expression by phosphorylating the coactivator transducer of regulated CREB activity (TORC2). Phosphorylation at Ser(587) of SIK2 diminishes its TORC2 phosphorylation activity. In 3T3-L1 white adipocytes, SIK2 downregulates lipogenic gene in response to nutritional stresses. To investigate the impact of SIK2 on the function of brown adipose tissue (BAT), we used T37i brown adipocytes, mice with diet-induced obesity, and SIK2 mutant (S587A) transgenic mice. When T37i adipocytes were treated with insulin, the levels of peroxisome proliferator-activated receptor-coactivator-1alpha (PGC-1alpha) and uncoupling protein-1 (UCP-1) mRNA were increased, and the induction was inhibited by overexpression of SIK2 (S587A) mutant or dominant-negative CREB. Insulin enhanced SIK2 phosphorylation at Ser(587), which was accompanied by decrease in phospho-TORC2. Similarly, the decrease in the level of SIK2 phosphorylation at Ser(587) was observed in the BAT of mice with diet-induced obesity, which was negatively correlated with TORC2 phosphorylation. To confirm the negative correlation between SIK2 phosphorylation at Ser(587) and TORC2 phosphorylation in BAT, SIK2 mutant (S587A) was overexpressed in adipose tissues by using the adipocyte fatty acid-binding protein 2 promoter. The expression of recombinant SIK2 (S587A) was restricted to BAT, and the levels of phospho-TORC2 were elevated in BAT of transgenic mice. Male transgenic mice developed high-fat diet-induced obesity, and their BAT expressed low levels of PGC-1alpha and UCP-1 mRNA, suggesting that SIK2-TORC2 cascade may be important for the regulation of PGC-1alpha and UCP-1 gene expression in insulin signaling in BAT.