MilliporeSigma
  • Digestive enzymes and stylet morphology of Deraeocoris nigritulus (Uhler)(Hemiptera: Miridae) reflect adaptations for predatory habits.
Digestive enzymes and stylet morphology of <I>Deraeocoris nigritulus</I> (Uhler)(Hemiptera: Miridae) reflect adaptations for predatory habits.
Annals of the Entomological Society of America (2003)
Boyd Jr, D. W.