MilliporeSigma
  • Asymmetric induction with β-cyclodextrin: cis-trans photoisomerization of diphenyl-cyclopropane and its derivatives.
Asymmetric induction with β-cyclodextrin: cis-trans photoisomerization of diphenyl-cyclopropane and its derivatives.
Proceedings of the Indian Academy of Sciences - Section A (2002)
Koodanjeri S, et al.