MilliporeSigma
EphA4 is a prognostic factor in gastric cancer.
BMC clinical pathology (2013-06-07)
Kohji Miyazaki, Mikito Inokuchi, Yoko Takagi, Keiji Kato, Kazuyuki Kojima, Kenichi Sugihara
ABSTRACT
Erythropoietin-producing hepatocellular (Eph) receptor, consisting of a family of receptor tyrosine kinases, plays critical roles in tumour development and is considered an attractive target for cancer therapy. Tumour samples were obtained from 222 patients with gastric adenocarcinoma who underwent gastrectomy. The expressions of EphA2, EphA4, and ephrinA1 were evaluated immunohistochemically. High expressions of EphA2, EphA4, and ephrinA1 significantly correlated with variables related to tumour progression, including the depth of invasion, metastatic lymph nodes, pathological stage, and distant metastasis or recurrent disease. High expressions of EphA2, EphA4, and ephrinA1 were significantly associated with poorer disease-specific survival (DSS; p < 0.001, p < 0.001, p = 0.026). On multivariate analysis, EphA4 was an independent prognostic factor of DSS (hazard ratio [HR], 2.3; 95% confidence interval [CI], 1.1-4.8; p = 0.028), and EphA2 tended to be a prognostic factor (HR, 2.4; 95% CI, 1.0-5.8; p = 0.050). In stage II and III cancer, EphA4 and EphA2 were both significantly associated with shorter survival (p = 0.007 and 0.019), but only EphA2 was an independent prognostic factor (HR, 2.6; 95% CI, 1.1-6.3; p = 0.039). EphA4 may play important roles in tumor progression and outcomes in patients with gastric cancer.