MilliporeSigma
  • Biomass conversion to high value chemicals: from furfural to chiral hydrofuroins in two steps.
Biomass conversion to high value chemicals: from furfural to chiral hydrofuroins in two steps.
Organic letters (2012-07-27)
Anzhelika Kabro, Eduardo C Escudero-Adán, Vladimir V Grushin, Piet W N M van Leeuwen
ABSTRACT
Catalytic asymmetric transfer hydrogenation of rac-furoin and furil produces hydrofuroin with up to 99% ee and 9:1 dr. This reaction provides an exceptionally easy access to optically active hydrofuroins in two straightforward steps from biomass-derived furfural (global production 200,000-300,000 t annually) using benzoin condensation.