MilliporeSigma
An improved fractionation system for pronase on CM-sephadex.
Canadian journal of biochemistry (1971-11-01)
J Jurásek, P Johnson, R W Olafson, L B Smillie
Sigma-Aldrich
Protease from Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder